missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals CHES, 99%
351.97 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
CHES is zwitterionic buffer useful in pH range 8.6 to 10.0.CHES is widely utilized as a buffer which is used for investigations on pH-dependent processes in enzymology. It has been shown to have an unusually high affinity for the iodoacetate binding site of liver alcohol dehydrogenase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
CHES is zwitterionic buffer useful in pH range 8.6 to 10.0.CHES is widely utilized as a buffer which is used for investigations on pH-dependent processes in enzymology. It has been shown to have an unusually high affinity for the iodoacetate binding site of liver alcohol dehydrogenase.
Solubility
Soluble in water.
Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
CHES is zwitterionic buffer useful in pH range 8.6 to 10.0.CHES is widely utilized as a buffer which is used for investigations on pH-dependent processes in enzymology. It has been shown to have an unusually high affinity for the iodoacetate binding site of liver alcohol dehydrogenase.
Solubility
Soluble in water.
Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | >300°C |
Quantity | 500 g |
Assay Percent Range | 0.99 |
Beilstein | 2967601 |
Merck Index | 14,2055 |
Solubility Information | Soluble in water. |
Formula Weight | 207.29 |
Percent Purity | 99% |
Chemical Name or Material | CHES |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'