missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chicken anti-Human IgG Fc Secondary Antibody, FITC
Chicken Polyclonal Secondary Antibody
Brand: Invitrogen™ SA172047
This item is not returnable.
View return policy
Description
SA1-72047 detects Human IgG Fc fragment in human samples. SA1-72047 has been successfully used in immunohistochemistry procedures. The SA1-72047 immunogen is from purified IgG from normal human serum.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Human IgG Fc | |
Polyclonal | |
FITC | |
Chicken | |
purified | |
RUO | |
Human | |
Secondary Antibody | |
IgY |
Immunohistochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.075% sodium azide | |
Purified IgG from normal human serum. | |
500 μg | |
Secondary | |
-20°C, store in dark | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Chicken anti-Human IgG Fc Secondary Antibody, FITC > 500 μg; FITC
Spot an opportunity for improvement?Share a Content Correction