Learn More
Chikungunya Virus Monoclonal Antibody (J31F), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA518182
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody is specific to Chikungunya virus and does not cross-react with other Alphaviruses Eastern Equine Encephalitis, Western Equine Encephalitis, Venezuelan Equine Encephalitis or other Arboviruses. Purity is >90%.
Chikungunya Virus Monoclonal antibody specifically detects Chikungunya Virus in Virus samples. It is validated for ELISA, Immunocytochemistry.Specifications
Chikungunya Virus | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2b |
ELISA, Immunocytochemistry | |
J31F | |
Unconjugated | |
CHIKV | |
Chikungunya virus native protein | |
100 μg | |
Primary | |
4° C | |
Liquid |