missing translation for 'onlineSavingsMsg'
Learn More

Logistik™ Child-Size Laboratory Coats

Product Code. 12813407
Click to view available options
Description:
Childrens Ages 10 to 12
Childrens Ages 7 to 9
Size:
Medium
Large
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Protects clothing whilst conducting experiments in the classroom. Logistik™ Child-Size Laboratory Coats features two lower patch pockets, one breast pocket, and concealed stud front.

  • Poly-cotton blend
  • Three pockets
  • Stud fasteners

Specifications

Product Type Children's Lab Coat
Size Large
Gender Unisex
Material Carbon, Polyester
Color White
Cuff Style Stud-fastened
Disposable Reusable
Sterility Non-sterile
No. of Pockets 3
Description Childrens Ages 10 to 12
For Use With (Application) Helps protect clothing whilst conducting experiments in the classroom
Includes With two lower patch pockets and one breast pocket and concealed stud front
Construction 65% Polyester, 35% Cotton
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'