missing translation for 'onlineSavingsMsg'
Learn More

Chloral hydrate, 98.5%, Thermo Scientific Chemicals

Product Code. 10013000
Click to view available options
Quantity:
50 g
500 g
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-17-0
Molecular Formula C2H3Cl3O2
Molecular Weight (g/mol) 165.39
MDL Number MFCD00044479
InChI Key RNFNDJAIBTYOQL-UHFFFAOYSA-N
IUPAC Name 2,2,2-trichloroethane-1,1-diol
SMILES OC(O)C(Cl)(Cl)Cl

Specifications

Melting Point 57.0°C
Boiling Point 97.0°C
Assay Percent Range 98.5%
Packaging Glass bottle
Linear Formula Cl3CCH(OH)2
Quantity 50 g
Merck Index 15, 2071
Solubility Information Solubility aq. soln.: 660 g/100 mL. Other solubilities: soluble in chloroform, alcohol and ether, freely soluble in acetone and methyl ethyl ketone, sparingly soluble in petroleum ether, benzene,, carbon tetrachloride, toluene and turpentine
Formula Weight 165.4
Chemical Name or Material Chloral hydrate
compliance-icons
Product Identifier
  • Chloral hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.