missing translation for 'onlineSavingsMsg'
Learn More

Chloramine-T trihydrate, 97+%

Product Code. p-3734178 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
1kg
250g
25g
5kg
This item is not returnable. View return policy

Product Code. 10695092

Brand: Thermo Scientific Acros 227850250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chloramine-T trihydrate, >-97%, C7H14ClNNaO5S, CAS Number-7080-50-4, 5kg, 97+%, White, 192 deg.C, 281.69, GHS Signal Word: Danger | CAS: 7080-50-4 | C7H13ClNNaO5S | 281.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7080-50-4
Molecular Formula C7H13ClNNaO5S
Molecular Weight (g/mol) 281.68
MDL Number MFCD00149066
InChI Key RNROOTWAJBXJBD-UHFFFAOYSA-M
PubChem CID 122197679
SMILES O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl

Specifications

Type Chloramine T Trihydrate
Physical Form Crystalline Powder
pH 8 to 10.3 (5% soln.)
Melting Point 170.0°C to 177.0°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Color White
Quantity 25 g
Flash Point 192°C
Assay Percent Range 97+%
Linear Formula CH3C6H4SO2N(Cl)Na·3H2O
Merck Index 15, 2075
IUPAC Name N-chloro-4-methylbenzenesulfonamide;sodium;trihydrate
Formula Weight 281.69
Percent Purity ≥97%
Chemical Name or Material Chloramine-T trihydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chloramine-T trihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.