missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Chloramphenicol, 98% Product Code.: 10368030

Thermo Scientific Chemicals Chloramphenicol, 98%

Product Code. 10368030
25 g, Glass Bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10368030

Brand: Thermo Scientific Chemicals 227920250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chloramphenicol, CAS # 56-75-7, is a preparation of the broad-spectrum, semisynthetic antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Chloramphenicol is an organochloride compound that contains a nitrobenzene ring
  • Chloramphenicol acts as a bacteriostatic by binding the 50S ribosomal subunit and preventing peptide formation
  • Stops peptidyltransferase activity by complexing to 50S subunit

Applications

  • Intended for molecular and microbiology applications such as antibiotic resistance gene testing and selective growth experiments and as a selection agent in bacterial cell culture
  • For research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-75-7
Molecular Formula C11H12Cl2N2O5
Molecular Weight (g/mol) 323.126
MDL Number MFCD00078159
InChI Key WIIZWVCIJKGZOK-RKDXNWHRSA-N
Synonym Chloromycetin, D(-)-threo-2, 2-Dichloro-N-[β-hydroxy-α-(hydroxy-methyl)-p-nitrophenylethyl]acetamide
PubChem CID 5959
ChEBI CHEBI:17698
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
SMILES C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]

Specifications

Type Chloramphenicol
Melting Point 149.0°C to 153.0°C
Packaging Glass Bottle
Specific Rotation + 18.50
Quantity 25 g
Assay Percent Range 98%
Solubility Information Solubility in water: 2.5g/L (25°C). Other solubilities: 150.8 mg/mL propylene glycol, soluble in methanol, ethanol, butanol, acetone and, ethyl acetate - slightly soluble in ether, insoluble in benzene and petroleum ether
Formula Weight 323.13
Percent Purity 98%
Chemical Name or Material Chloramphenicol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468968 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number227921000,227920250,227925000
Lot NumberA0463972 Certificate TypeCertificate of Analysis Date03/07/2024 Catalog Number227921000,227920250,227925000
Lot NumberA0458368 Certificate TypeCertificate of Analysis Date01/12/2023 Catalog Number227921000,227920250,227925000
Lot NumberA0456356 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number227921000,227920250,227925000
Lot NumberA0451580 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number227921000,227920250,227925000
compliance-icons
Product Identifier
  • Chloramphenicol
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Chloramphenicol, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.