missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloride ISE Standard Solution, Mettler Toledo™
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Physical Form | Liquid |
Packaging | 500 mL |
Color | Colorless |
Quantity | 500 mL |
Description | ISE standard Cl 1000 mg/L, 500mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction