missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloride Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical A 013 250
This item is not returnable.
View return policy
Specifications
Chloride | |
Bottle | |
Single-component Standard Solution | |
H2O |
1g/L | |
250 mL | |
Cl- | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction