Learn More
Chlorinated Disinfection By-Products, Solvents and Trihalomethanes Combination Mix, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep 5511-A
Additional Details : CAS Number : 3252-43-5 Weight : 0.00100kg Transport : UN number : 1090 Chem class : 3 Pack group : II
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
1,1,1-Trichloroethane | 71-55-6 | 2000μg/mL |
1,1-Dichloroacetone | 513-88-2 | 2000μg/mL |
1,2-Dibromo-3-chloropropane | 96-12-8 | 2000μg/mL |
1,2-Dibromoethane | 106-93-4 | 2000μg/mL |
Bromodichloromethane | 75-27-4 | 2000μg/mL |
Bromoform | 75-25-2 | 2000μg/mL |
Carbon tetrachloride | 56-23-5 | 2000μg/mL |
Chloroform | 67-66-3 | 2000μg/mL |
Chloropicrin | 76-06-2 | 2000μg/mL |
Dibromoacetonitrile | 3252-43-5 | 2000μg/mL |
Dibromochloromethane | 124-48-1 | 2000μg/mL |
Dichloroacetonitrile | 3018-12-0 | 2000μg/mL |
Tetrachloroethene | 127-18-4 | 2000μg/mL |
Trichloroacetonitrile | 545-06-2 | 2000μg/mL |
Trichloroethene | 79-01-6 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Chlorinated Disinfection By-Products Mix | |
1 mL | |
71-55-6,96-12-8,106-93-4,75-25-2,56-23-5 | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
For US EPA Method 550 | |
Acetone |
2mL amber prescored glass ampule | |
0.2 | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
2000μg/mL | |
1,1,1-Trichloroethane |
Safety and Handling
- Chlorinated Disinfection By-Products Mix
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetone, carbon tetrachloride, chloroform, bromoform, bromodichloromethane, trichloronitromethane, t
Product intended for laboratory use only