missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chlorine Test Paper

Product Code. 11376514 Shop All Macherey-Nagel Products
Click to view available options
Color Indicator:
White to Dark Blue
Unit Size:
5m
This item is not returnable. View return policy

Product Code. 11376514

Brand: Macherey-Nagel™ 90709

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allows for the semi-quantitative determination of 50-200ppm chlorine. Macherey Nagel™ Chlorine Test Paper is non-toxic and safe.

  • White paper turns blue-violet in the presence of chlorine
  • Particularly suitable for a range of applications in disinfectant solutions

Specifications

Color Indicator White to Dark Blue
Format Reel
For Use With (Application) Determination of Chlorine
Graduations 0, 10, 50, 100, 200
Quantity 1 Each
Type pH Paper
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.