missing translation for 'onlineSavingsMsg'
Learn More
Chloro(1,5-cyclooctadiene)iridium(I) dimer, Ir 57.2%, Thermo Scientific Chemicals Product Code.: 11327537

Chloro(1,5-cyclooctadiene)iridium(I) dimer, Ir 57.2%, Thermo Scientific Chemicals

Product Code. 11327537
0.25 g
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy

Product Code. 11327537

Brand: Thermo Scientific Alfa Aesar 012749.01

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chloro(1,5-cyclooctadiene)iridium(I) dimer is widely used as a precursor to other iridium complexes, which finds application in homogeneous catalysis like carbonylation, hydrosilylation, hydrofomylation, asymmetric allylic substitutions, metathesis and chiral catalysis reactions. It is involved in the preparation of Crabtree's catalyst, which is used for hydrogenation and hydrogen-transfer reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12112-67-3
Molecular Formula C16H24Cl2Ir2
Molecular Weight (g/mol) 671.70
MDL Number MFCD00012414
InChI Key ZFOUDQNHNLDNLD-MIXQCLKLSA-L
Synonym chloro 1,5-cyclooctadiene iridium i dimer, bis 1,5-cyclooctadiene diiridium i dichloride, 1z,5z-cycloocta-1,5-diene; iridium; dichloride, iridium, di-.mu.-chlorobis 1,2,5,6-.eta.-1,5-cyclooctadiene di, ir2cl2 cod 2, chloro-1,5-cyclooctadiene iridium i dimer, 1z,5z-cycloocta-1,5-diene; iridium 1+
PubChem CID 6436381
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;iridium;dichloride
SMILES [Cl-].[Cl-].[Ir+].[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 190°C (decomposition)
Color White
Odor Odorless
Quantity 0.25 g
Solubility Information Soluble in chloroform and toluene. Slightly soluble in acetone and alcohol. Insoluble in water.
Formula Weight 671.71
Physical Form Crystalline Powder
Chemical Name or Material Chloro(1, 5-cyclooctadiene)iridium(I) dimer

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX25E029 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog Number012749.03,012749.01,012749.06
Lot NumberS06K018 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number012749.03,012749.01,012749.06
Lot NumberY28J010 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number012749.03,012749.01,012749.06
Lot NumberU09J027 Certificate TypeCertificate of Analysis Date16/08/2023 Catalog Number012749.03,012749.01,012749.06
Lot NumberY16I029 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number012749.03,012749.01,012749.06,012755.03
compliance-icons
Product Identifier
  • Chloro(1,5-cyclooctadiene)iridium(I) dimer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Chloro(1,5-cyclooctadiene)iridium(I) dimer

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chloro(1,5-cyclooctadiene)iridium(I) dimer, Ir 57.2%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.