missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 143°C to 147°C |
Color | Brown-Yellow |
Quantity | 1 g |
UN Number | UN3395 |
Synonym | Pentamethylcyclopentadienyl(1,5-cyclooctadiene)ruthenium(II) chloride; RuCl(cod)Cp* |
Formula Weight | 379.94 |
Physical Form | Crystalline Solid |
Chemical Name or Material | Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II) |
Safety and Handling

Product Identifier
- Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
- Danger
- Substance which\, in contact with water\, emits flammable gases Category 2
- H261-In contact with water releases flammable gases.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : Chloro(pentamethylcyclopentadienyl)(cyclo octadiene) ruthenium(II)
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction