Learn More
Chloro(1,5-cyclooctadiene)rhodium(I) dimer, Thermo Scientific Chemicals
Description
It is a widely used precursor to homogeneous catalysts. This is a chiral catalyst capable of asymmetrically hydrogenating certain prochiral alkenes. Chloro(1,5-cyclooctadiene)rhodium(I) dimer is also used in the synthesis of other metal ligands for use in catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 243°C (decomposition) |
Color | Yellow to Orange |
Odor | Odorless |
Quantity | 250 mg |
Sensitivity | Air sensitive; Hygroscopic |
Solubility Information | Soluble in chloroform,dichloromethane,and methanol; Sparingly soluble in most common solvents. |
Formula Weight | 493.08 |
Physical Form | Crystalline |
Chemical Name or Material | Chloro(1,5-cyclooctadiene)rhodium(I) dimer |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.