missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloro(crotyl)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
Description
Chloro(crotyl)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II) acts as a ligand and chiral catalyst in synthetic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 1252598-33-6 |
Color | Pink |
Molecular Formula | C52H54ClFePPd |
MDL Number | MFCD25372545 |
Quantity | 1 g |
Synonym | pdcl crotyl qphos, chloro crotyl 1,2,3,4,5-pentaphenyl-1'-di-tert-butylphosphino ferrocene palladium ii |
Solubility Information | Insoluble in water. |
InChI Key | ZDJQQIXNGDRDQM-UHFFFAOYSA-M |
SMILES | Cl[Pd++].C\C=C/C.CC(C)(C)P(c1cccc1[Fe])C(C)(C)C.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
IUPAC Name | λ²-iron(2+) 3-(di-tert-butylphosphanyl)cyclopenta-2,4-dien-1-ide but-2-en-1-ide chloropalladiumylium pentaphenylcyclopenta-2,4-dien-1-ide |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chloro(crotyl)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction