Learn More
Chloro(methyl)diphenylsilane, 97%, Thermo Scientific Chemicals
£27.70 - £81.30
Chemical Identifiers
CAS | 144-79-6 |
---|---|
Molecular Formula | C13H13ClSi |
Molecular Weight (g/mol) | 232.78 |
MDL Number | MFCD00000498 |
InChI Key | OJZNZOXALZKPEA-UHFFFAOYSA-N |
Synonym | chloro methyl diphenylsilane, methyldiphenylchlorosilane, chloromethyldiphenylsilane, silane, chloromethyldiphenyl, chlorodiphenylmethylsilane, diphenylmethylchlorosilane, methyldiphenylsilyl chloride, dpmscl, chloro-methyl-diphenyl-silane, benzene, 1,1'-chloromethylsilylene bis |
PubChem CID | 67349 |
IUPAC Name | chloro-methyl-diphenylsilane |
SMILES | C[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11376816
|
Thermo Scientific Alfa Aesar
L04162.09 |
10 g |
£27.70
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386816
|
Thermo Scientific Alfa Aesar
L04162.18 |
50 g |
£81.30
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chloro(methyl)diphenylsilane is used in agrochemical, pharmaceutical and dyestuff field .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChloro(methyl)diphenylsilane is used in agrochemical, pharmaceutical and dyestuff field .
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from strong bases. Incompatible with moisture and bases.
Chemical Identifiers
144-79-6 | |
232.78 | |
OJZNZOXALZKPEA-UHFFFAOYSA-N | |
67349 | |
C[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1 |
C13H13ClSi | |
MFCD00000498 | |
chloro methyl diphenylsilane, methyldiphenylchlorosilane, chloromethyldiphenylsilane, silane, chloromethyldiphenyl, chlorodiphenylmethylsilane, diphenylmethylchlorosilane, methyldiphenylsilyl chloride, dpmscl, chloro-methyl-diphenyl-silane, benzene, 1,1'-chloromethylsilylene bis | |
chloro-methyl-diphenylsilane |
Specifications
294°C to 295°C | |
C13H13ClSi | |
UN2987 | |
141°C (285°F) | |
Reacts with water. | |
C[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1 | |
232.78 | |
232.79 | |
Irritating | |
Moisture sensitive | |
Chloro(methyl)diphenylsilane |
144-79-6 | |
MFCD00000498 | |
2937445 | |
chloro methyl diphenylsilane, methyldiphenylchlorosilane, chloromethyldiphenylsilane, silane, chloromethyldiphenyl, chlorodiphenylmethylsilane, diphenylmethylchlorosilane, methyldiphenylsilyl chloride, dpmscl, chloro-methyl-diphenyl-silane, benzene, 1,1'-chloromethylsilylene bis | |
OJZNZOXALZKPEA-UHFFFAOYSA-N | |
chloro-methyl-diphenylsilane | |
67349 | |
97% | |
1.574 | |
1.104 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 205-639-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only