missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloro(triphenylphosphine)gold(I), Premion™, 99.99% (metals basis), Au 39.3% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
1 g
5 g
Description
Chloro(triphenylphosphine)gold(I) is a common reagent in gold chemistry. It catalyzes certain rearrangement reactions in organic synthesis. It is also used to prepare 1-methylthyminato-N3-triphenylphosphinegold(I) by reacting with 1-methylthymine. It acts as a precursor for the synthesis of gold(I) and gold(III) organometallic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Yellow |
Assay Percent Range | 99.99% (Metal basis, Au 39.3% min.) |
Odor | Odorless |
Quantity | 0.5 g |
Solubility Information | Soluble in methylene chloride,acetonitrile,benzene and acetone. Insoluble in water and ethanol. |
Formula Weight | 494.71 |
Physical Form | Powder |
Chemical Name or Material | Chloro(triphenylphosphine)gold(I) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'