missing translation for 'onlineSavingsMsg'
Learn More
Chloroacetic acid, 99%, flakes, Thermo Scientific Chemicals Product Code.: 10335790

Chloroacetic acid, 99%, flakes, Thermo Scientific Chemicals

Product Code. 10335790
250 g, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
4 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10335790

Brand: Thermo Scientific Acros 108512500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-11-8
Molecular Formula C2H3ClO2
Molecular Weight (g/mol) 94.5
MDL Number MFCD00002683
InChI Key FOCAUTSVDIKZOP-UHFFFAOYSA-N
Synonym chloroacetic acid, monochloroacetic acid, chloracetic acid, acetic acid, chloro, acide chloracetique, chloroethanoic acid, monochloroethanoic acid, monochloracetic acid, monochloorazijnzuur, monochloressigsaeureShow MoreShow Less
PubChem CID 300
ChEBI CHEBI:27869
IUPAC Name 2-chloroacetic acid
SMILES C(C(=O)O)Cl

Specifications

Decomposition Information 250°C
Melting Point 61.0°C to 63.0°C
Color White
Density 1.5800g/mL
Boiling Point 189.0°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula ClCH2CO2H
Quantity 250 g
Beilstein 02, 194
Merck Index 15, 2112
Specific Gravity 1.58
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, chloroform, ether
Viscosity 1.29 Pas (100°C)
Formula Weight 94.5
Vapor Pressure 0.75 mmHg at 20°C
Percent Purity 99%
Physical Form Crystalline Flakes
Chemical Name or Material Chloroacetic acid, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464757 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog Number108510025,108512500,108510040,108510010
Lot NumberA0463040 Certificate TypeCertificate of Analysis Date27/05/2024 Catalog Number108510025,108512500,108510040,108510010
Lot NumberA0463039 Certificate TypeCertificate of Analysis Date24/05/2024 Catalog Number108510025,108512500,108510040,108510010
Lot NumberA0459693 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog Number108510025,108512500,108510040,108510010
Lot NumberA0457212 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number108510025,108512500,108510040,108510010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chloroacetic acid
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chloroacetic acid, 99%, flakes, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.