missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobenzene, 99+%, for spectroscopy, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
Specifications
Melting Point | -45.0°C |
Absorbance | 0.01 max. at 360nm, 0.07 max. at 300nm, 0.3 max. at 290nm, 1 max. at 287nm |
Density | 1.1075g/mL |
Boiling Point | 131.0°C |
Quantity | 500 mL |
Flash Point | 23°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.5230 to 1.5250 |
Assay Percent Range | 99+% |
Show More |
Safety and Handling
Product Identifier
- Chlorobenzene
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'