missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobenzene, Certified AR for Analysis, Fisher Chemical™
63.08 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Plastic-coated Glass Bottle
Specifications
Melting Point | -45°C |
Boiling Point | 131°C |
Quantity | 2.5 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 12mbar at 20°C |
Viscosity | 0.8 mPaS at 20°C |
Formula Weight | 112.56g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'