missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobenzene, CHROMASOLV™, for HPLC, 99.9%, Honeywell Riedel-de Haen
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Melting Point | -45°C (literature) |
Absorbance | max. 1.0 at 288 nm, max. 0.01 at 400 nm, max. 0.01 at 360 nm, max. 0.04 at 325 nm, max. 0.05 at 300 nm, max. 0.40 at 290 nm |
CAS | 108-90-7 |
CAS Max % | 100.0000% |
Density | 1.106 g/mL at 25 °C(lit.) |
Boiling Point | 130°C to 132°C |
Quantity | 1 L |
Flash Point | 28°C |
Packaging | Glass Bottle |
Molecular Formula | C6H5Cl |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'