missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobenzene-d{5}, 99% (Isotopic), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Chlorobenzene-d5 is a halogenated D5 chlorobenzene for proteomics research. It is a precursor to deuterated dyes and rubbers. It is used as a high-boiling solvent in industrial applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Liquid |
Density | 1.157 g/mL |
Flash Point | 24°C (75°F) |
Odor | Aromatic |
Refractive Index | 1.522 |
Quantity | 5 g |
Boiling Point | 130°C |
UN Number | UN1134 |
Beilstein | 1866335 |
Solubility Information | Not miscible with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'