missing translation for 'onlineSavingsMsg'
Learn More
Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine, 97%, maycontain suspended amine hydrochloride crystals, Thermo Scientific Chemicals Product Code.: 11495620

Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine, 97%, maycontain suspended amine hydrochloride crystals, Thermo Scientific Chemicals

Product Code. 11495620
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11495620

Brand: Thermo Scientific Alfa Aesar H25883.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine is used as a reactant for the synthesis of chiral phosphine-aminophosphine ligands for rhodium-catalyzed asymmetric hydrogenation, ligands for palladium-catalyzed stereoselective allylation reactions, enantioselective hydrogenations, Josiphos analog as catalyst for asymmetric hydrogenation, ligands used in asymmetric hydrovinylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine is used as a reactant for the synthesis of chiral phosphine-aminophosphine ligands for rhodium-catalyzed asymmetric hydrogenation, ligands for palladium-catalyzed stereoselective allylation reactions, enantioselective hydrogenations, Josiphos analog as catalyst for asymmetric hydrogenation, ligands used in asymmetric hydrovinylation reactions.

Solubility
Reacts with water.

Notes
Air & moisture sensitive. Store away from oxidizing agents, bases, air, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142421-57-6
Molecular Formula C16H6ClF12P
Molecular Weight (g/mol) 492.629
MDL Number MFCD01630852
InChI Key DFZQEHBNAJGDCT-UHFFFAOYSA-N
Synonym bis 3,5-di trifluoromethyl phenyl chlorophosphine, phosphinous chloride, bis 3,5-bis trifluoromethyl phenyl, acmc-20alnu, bis 3,5-bis trifluoromethyl phenyl chlorophosphine, bis 3,5-bis trifluoromethyl phenyl-chlorophosphane, bis 3,5-bis trifluoromethyl phenyl-chlorophosphine, bis 3,5-bis trifluoromethyl phenyl chloro phosphane, p,p-bis 3,5-bis trifluoromethyl phenyl phosphinous chloride, phosphinous chloride,p,p-bis 3,5-bis trifluoromethyl phenylShow MoreShow Less
PubChem CID 5104965
IUPAC Name bis[3,5-bis(trifluoromethyl)phenyl]-chlorophosphane
SMILES C1=C(C=C(C=C1C(F)(F)F)P(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)Cl)C(F)(F)F

Specifications

Quantity 1 g
UN Number UN3265
Sensitivity Air and moisture sensitive; May form a precipitate
Solubility Information Reacts with water.
Formula Weight 492.63
Percent Purity ≥98%
Assay may contain suspended dimethylamine hydrochloride crystals
Chemical Name or Material Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10245595 Certificate TypeCertificate of Analysis Date01/02/2023 Catalog NumberH25883.03,H25883.06
Lot Number10189284 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25883.03,H25883.06
Lot Number10226879 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25883.03,H25883.06
Lot Number10183163 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25883.03,H25883.06
Lot Number10215355 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25883.03,H25883.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine, 97%, maycontain suspended amine hydrochloride crystals, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.