missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis(4-fluorophenyl)phosphine, 98%, Thermo Scientific Chemicals

Product Code. 15493567
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

In chemical research. Used as an interphasic transfer catalyst, a polymerization catalyst, and a trans-halogenation catalyst. As a precursor to fire-retardant materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In chemical research. Used as an interphasic transfer catalyst, a polymerization catalyst, and a trans-halogenation catalyst. As a precursor to fire-retardant materials.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23039-97-6
Molecular Formula C12H8ClF2P
Molecular Weight (g/mol) 256.617
MDL Number MFCD04972302
InChI Key PYFWYPBHSZATST-UHFFFAOYSA-N
Synonym bis 4-fluorophenyl chlorophosphine, chlorobis 4-fluorophenyl phosphine, chloro-bis 4-fluorophenyl phosphane, chlorobis 4-fluorophenyl phosphane, acmc-20alnm, bis-4-fluorophenyl chlorophosphine, phosphinous chloride,p,p-bis 4-fluorophenyl
PubChem CID 4379931
IUPAC Name chloro-bis(4-fluorophenyl)phosphane
SMILES C1=CC(=CC=C1F)P(C2=CC=C(C=C2)F)Cl

Specifications

Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 256.62
Percent Purity 98%
Chemical Name or Material Chlorobis(4-fluorophenyl)phosphine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'