missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethylphenylsilane, 97+%, Thermo Scientific Chemicals

£31.30 - £338.00

Chemical Identifiers

CAS 768-33-2
Molecular Formula C8H11ClSi
Molecular Weight (g/mol) 170.71
MDL Number MFCD00000499
InChI Key KWYZNESIGBQHJK-UHFFFAOYSA-N
Synonym chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
PubChem CID 13029
IUPAC Name chloro-dimethyl-phenylsilane
SMILES C[Si](C)(Cl)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11457537
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15638.06
5 g
£31.30
5g
Please to purchase this item. Need a web account? Register with us today!
11467537
View Documents
Thermo Scientific Alfa Aesar
A15638.14
25 g
£85.30
25g
Please to purchase this item. Need a web account? Register with us today!
11477537
View Documents
Thermo Scientific Alfa Aesar
A15638.22
100 g
£338.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Chlorodimethylphenylsilane is used in the synthesis of enantioenriched allenylsilanes through an ortho-ester Claisen rearrangement of chiral and silylpropargylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodimethylphenylsilane is used in the synthesis of enantioenriched allenylsilanes through an ortho-ester Claisen rearrangement of chiral and silylpropargylic alcohols.

Solubility
Miscible with tetrahydrofuran, ether and hexane.

Notes
Incompatible with strong oxidizing agents, acids, bases, and moisture.
Specifications

Chemical Identifiers

768-33-2
170.71
KWYZNESIGBQHJK-UHFFFAOYSA-N
13029
C[Si](C)(Cl)C1=CC=CC=C1
C8H11ClSi
MFCD00000499
chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
chloro-dimethyl-phenylsilane

Specifications

192°C to 193°C
C8H11ClSi
UN2987
72°C (161°F)
Miscible with tetrahydrofuran,ether and hexane.
C[Si](C)(Cl)C1=CC=CC=C1
170.71
170.72
1.509
1.032
768-33-2
MFCD00000499
606292
chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
KWYZNESIGBQHJK-UHFFFAOYSA-N
chloro-dimethyl-phenylsilane
13029
≥97%
Moisture sensitive
Chlorodimethylphenylsilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 212-193-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only