missing translation for 'onlineSavingsMsg'
Learn More

chlorodimethylsilane, 96%, Thermo Scientific Chemicals

Product Code. 10711935
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-35-9
Molecular Formula C2H6ClSi
Molecular Weight (g/mol) 93.61
MDL Number MFCD00000495
InChI Key QABCGOSYZHCPGN-UHFFFAOYSA-N
Synonym chlorodimethylsilane, dimethylchlorosilane, dmcs, silane, chlorodimethyl, unii-xr7e4f5838, chloro dimethyl silicon, chlorodimethyl-silan, dimethylmono-chlorosilane, chlorodimethylsilyl radical
PubChem CID 6327132
IUPAC Name chloro(dimethyl)silicon
SMILES C[Si](C)Cl

Specifications

Melting Point -111°C
Color Colorless
Density 0.8500g/mL
Boiling Point 34.6°C
Flash Point -28°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula (CH3)2SiHCl
Refractive Index 1.3820 to 1.384
Quantity 250 g
Fieser 13,73
Specific Gravity 0.85
Solubility Information Solubility in water: reacts
Formula Weight 94.62
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material chlorodimethylsilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 A
  • Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H261-In contact with water releases flammable gases.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'