missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, ≥99.5%, Honeywell™
Click to view available options
Quantity:
25 mL
4 x 25 mL
500 mL
1 L
2.5 L
6 x 500 mL
Packaging:
Glass bottle
Descripción
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Especificaciones
Especificaciones
Melting Point | -61.3°C |
Color | Colorless |
Density | 1.492g/mL at 25°C |
Boiling Point | 61°C (1013 hPa) |
Odor | Sweet |
Packaging | Glass bottle |
Quantity | 6 x 500 mL |
UN Number | UN1888 |
Beilstein | 1731042 |
IUPAC Name | chloroform |
Mostrar más |
Seguridad y manipulación
Nombre del producto
- Chloroform
- Danger
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'