missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, 99.9%, for residue analysis, ECD tested for pest anal., residue <0., Thermo Scientific Chemicals

Product Code. 10237380
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Metal Drum
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For residue analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3, 513-35-9
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name chloroform
SMILES ClC(Cl)Cl

Specifications

Melting Point -63.0°C
Density 1.4800g/mL
Boiling Point 61.0°C
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 99.9%
Residue after Evaporation 0.0005% max.
Specific Gravity 1.48
Linear Formula CHCl3
Viscosity 0.56 mPa.s (20°C)
Water 0.01% max. (Coulometric)
Fieser 01,130; 08,92; 15,84
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in acetone.
Formula Weight 119.38
Percent Purity 99.9%
Grade Residue Analysis
Chemical Name or Material Chloroform, ECD Tested, For Pesticide Analysis, Residue <0.0005%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroform, stabilized with amylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H372-Causes damage to organs.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.