missing translation for 'onlineSavingsMsg'
Learn More
Chloroform, 99+%, extra pure, stabilized with ethanol Product Code.: 15771819

Chloroform, 99+%, extra pure, stabilized with ethanol

Product Code. 15771819
10 L, Plastic Drum
Click to view available options
Quantity:
10 L
Packaging:
Plastic Drum
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15771819

Brand: Thermo Scientific Acros 158210100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 67-66-3 | CHCl3 | 119.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 67-66-3, 64-17-5
Decomposition Information 290°C
Density 1.4800g/mL
Quantity 10 L
Packaging Plastic Drum
Physical Form Liquid
Assay Percent Range 99+%
Residue after Evaporation 0.002% max.
Molecular Formula CHCl3
Linear Formula CHCl3
MDL Number MFCD00000826
Vapor Pressure Vapor Pressure: 213mbar at 20°C
Beilstein 01, 61
Water 0.02% max. (K.F.)
Fieser 01,130; 08,92; 15,84
Merck Index 15, 2142
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES ClC(Cl)Cl
IUPAC Name chloroform
Molecular Weight (g/mol) 119.37
PubChem CID 6212
ChEBI CHEBI:35255
Formula Weight 119.38
Percent Purity ≥99.0% (excluding stabilizer)
Grade Extra Pure
Chemical Name or Material Chloroform
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465855 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog Number158210050,158210025,158210100,158210010,158210250
Lot NumberA0396057 Certificate TypeCertificate of Analysis Date24/04/2024 Catalog Number158210050,158210025,158210100,158210010,158210250
Lot NumberA0444412 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog Number158210050,158210025,158210100,158210010,158210250
Lot NumberA0457638 Certificate TypeCertificate of Analysis Date16/11/2023 Catalog Number158210050,158210025,158210100,158210010,158210250
Lot NumberB0541096A Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number158210050,158210025,158210100,158210010,158210250
compliance-icons compliance-icons
Product Identifier
  • Chloroform, stabilized with ethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
  • Carcinogenicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H336-May cause drowsiness or dizziness.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chloroform, 99+%, extra pure, stabilized with ethanol >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.