missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, ACS Reagent, for determinations with dithizone, 99.0-99.4%, Honeywell Riedel-de Haen
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Melting Point | -63°C (literature) |
CAS | 67-66-3, 64-17-5 |
CAS Min % | ≥90.0000%, ≥0.1000% |
CAS Max % | ≤100.0000%, <1.0000% |
Molecular Formula | CHCl3 |
Linear Formula | CHCl3 |
MDL Number | MFCD00000826 |
UN Number | UN1888 |
Beilstein | 1731042 |
Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'