Learn More
Chloroform (Approx. 0.75% Ethanol as Preservative/Molecular Biology), Fisher BioReagents
Brand: Fisher Bioreagents BP1145-1
Code : 32
Additional Details : CAS Number : 67-66-3 Weight : 2.91776kg Transport : UN number : 1888 Chem class : 6.1 Pack group : III
Description
Glass bottle; 1L
This solvent is used in Phenol/Chloroform extractions to remove proteins from DNA or RNA samples.EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Chloroform | |
Approx. 0.75% Ethanol as Preservative | |
0.13 max. at 254nm, 0.28 max. at 250nm, 0.8 max. at 245nm, 1 max. at 244nm | |
290°C | |
61°C | |
Glass Bottle | |
1.4420 to 1.4450 (at 25°C) | |
≥99 % (by GC) | |
MFCD00000826 | |
0.56 mPa/s at 20°C | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 | |
≥99% |
-63°C | |
67-66-3,64-17-5 | |
Colorless | |
1.480g/cm³ | |
1 L | |
Liquid | |
2ppm max. | |
CHCl3 | |
213mbar at 20°C | |
0.02% max. | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
chloroform | |
6212 | |
119.38 | |
Molecular Biology |
Safety and Handling
- Chloroform, stabilized with ethanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H372-Causes damage to organs.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .