missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, Biotech Grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer, Honeywell Burdick & Jackson™
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass Bottle
Description
- Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
- As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
- Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
Specifications
Melting Point | -63°C (literature) |
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000% |
Density | 1.48 g/mL at 25 °C; 1.492 g/mL at 25 °C(lit.) |
Boiling Point | 61°C |
Quantity | 1 L |
Packaging | Glass Bottle |
Linear Formula | CH3CH2OH |
UN Number | UN1888 |
Beilstein | 1731042 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'