missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, CHROMASOLV™, for HPLC, contains 0.5-1.0% ethanol as stabilizer, ≥99.8%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass Bottle
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
Melting Point | -63°C (literature) |
Absorbance | max. 0.01 at 400 nm, max. 0.01 at 290 nm, max. 0.02 at 270 nm, max. 0.15 at 260 nm, max. 0.15 at 255 nm, max. 1.00 at 245 nm |
CAS Min % | ≥90.0000%, ≥0.1000% |
CAS Max % | ≤100.0000%, <1.0000% |
Linear Formula | H2SO4 |
UN Number | UN1888 |
Beilstein | 1731042 |
IUPAC Name | trichloromethane |
Grade | HPLC |
Density | 1.48 g/mL at 25 °C; 1.492 g/mL at 25 °C(lit.) |
Show More |
Safety and Handling
Product Identifier
- Chloroform
- Danger
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
- Reproductive toxicity Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'