missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, CHROMASOLV™, for HPLC, contains 0.5-1.0% ethanol as stabilizer, ≥99.8%, Honeywell Riedel-de Haën™

CHROMASOLV™, for HPLC, contains 0.5-1.0% ethanol as stabilizer,≥99.8%

£33.75 - £106.45

Chemical Identifiers

CAS 67-66-3, 64-17-5
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15695130
View Documents
Honeywell-Chromasolv
366927-100ML
100 mL Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15605140
View Documents
Honeywell-Chromasolv
366927-1L
1 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15615140
View Documents Promotion Details
Honeywell-Chromasolv
366927-2.5L
2.5 L Glass Bottle
£106.45
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
Specifications

Chemical Identifiers

67-66-3, 64-17-5
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
ClC(Cl)Cl
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255

Specifications

-63°C (literature)
67-66-3, 64-17-5
≤100.0000%, <1.0000%
60°C to 62°C
H2SO4
UN1888
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
ClC(Cl)Cl
119.37
CHEBI:35255
Chloroform, CHROMASOLV™, for HPLC, contains 0.5-1.0% ethanol as stabilizer, ≥99.8%
max. 0.01 at 400 nm, max. 0.01 at 290 nm, max. 0.02 at 270 nm, max. 0.15 at 260 nm, max. 0.15 at 255 nm, max. 1.00 at 245 nm
≥90.0000%, ≥0.1000%
1.48 g/mL at 25 °C; 1.492 g/mL at 25 °C(lit.)
CHCl3
MFCD00000826
1731042
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane
6212
HPLC
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 720 days from date of manufacture

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Chloroform

Recommended Storage : Room Temp

Product Identifier
  • Chloroform

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
  • Reproductive toxicity Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.