missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, Contains ethanol as stabilizer, meets analytical spec. of DAB9, BP, 99-99.4% (GC), Honeywell Riedel-de Haën™
Contains ethanol as stabilizer, meets analytical specification of DAB9, BP, 99-99.4% (GC)
£47.07 - £5538.82
Chemical Identifiers
CAS | 67-66-3, 64-17-5 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 119.37 |
MDL Number | MFCD00000826 |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
PubChem CID | 6212 |
ChEBI | CHEBI:35255 |
SMILES | ClC(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15672560
|
Honeywell-Riedel-de Haen
24216-1L |
1 L |
£47.07
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15682560
|
Honeywell-Riedel-de Haen
24216-2.5L |
2.5 L |
£72.85
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15622570
|
Honeywell-Riedel-de Haen
24216-6X1L |
6 x 1 L |
£228.87
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15612570
|
Honeywell-Riedel-de Haen
24216-4X2.5L |
4 x 2.5 L |
£243.99
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15602570
|
Honeywell-Riedel-de Haen
24216-40KG |
40 kg | N/A | ||||||
15692560
|
Honeywell Riedel-de Haën
24216-250KG |
250 kg |
£5538.82
250kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Chemical Identifiers
67-66-3, 64-17-5 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
ClC(Cl)Cl |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
CHEBI:35255 |
Specifications
-63°C (literature) | |
≥90.0000%, ≥0.1000% | |
1.479-1.489 g/mL at 20 °C; 1.492 g/mL at 25 °C(lit.) | |
Glass Bottle | |
CHCl3 | |
160mmHg (20°C) | |
1731042 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
trichloromethane | |
6212 | |
119.38g/mol | |
Reagents Pharmacopeia |
67-66-3, 64-17-5 | |
≤100.0000%, <1.0000% | |
60°C to 62°C | |
CHCl3 | |
MFCD00000826 | |
UN1888 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 | |
99 to 99.4% (GC) | |
Chloroform, Contains ethanol as stabilizer, meets analytical specification of DAB9, BP, 99-99.4% (GC) |
Safety and Handling
P260-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338-P403 + P233
H302-H315-H319-H331-H336-H351-H361d-H372
Danger
missing translation for 'shelfLife' : 1080 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Chloroform
missing translation for 'einecsNumber' : 200-663-8
missing translation for 'rtecsNumber' : FS9100000
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Chloroform
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title