Learn More
Chloroform-d, for NMR, 99.8% atom D, contains 1 v/v% TMS, Thermo Scientific Chemicals
£23.22 - £140.70
Chemical Identifiers
CAS | 865-49-6 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 120.375 |
MDL Number | MFCD00000827 |
InChI Key | HEDRZPFGACZZDS-MICDWDOJSA-N |
Synonym | chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero |
PubChem CID | 71583 |
ChEBI | CHEBI:85365 |
IUPAC Name | trichloro(deuterio)methane |
SMILES | C(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10215790
|
Thermo Scientific Acros
166260250 |
25 mL | Glass Bottle |
£23.22
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10551611
|
Thermo Scientific Acros
166260500 |
50 mL | Glass Bottle |
£40.70
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10418910
|
Thermo Scientific Acros
166261000 |
100 mL | Glass Bottle |
£71.80
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10347250
|
Thermo Scientific Acros
166262500 |
250 mL | Glass Bottle |
£140.70
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
865-49-6 | |
120.375 | |
HEDRZPFGACZZDS-MICDWDOJSA-N | |
71583 | |
trichloro(deuterio)methane |
CHCl3 | |
MFCD00000827 | |
chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero | |
CHEBI:85365 | |
C(Cl)(Cl)Cl |
Specifications
Crystals or Crystalline Powder | |
1.5000g/mL | |
0.02% max. (Coulometric) | |
-64.0°C | |
CHCl3 | |
MFCD00000827 | |
chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero | |
C(Cl)(Cl)Cl | |
120.375 | |
CHEBI:85365 | |
NMR |
White | |
99.75% min. | |
865-49-6 | |
60.0°C | |
CDCl3 | |
01, IV, 54 | |
HEDRZPFGACZZDS-MICDWDOJSA-N | |
trichloro(deuterio)methane | |
71583 | |
120.39 | |
Chloroform-d, 99.8 atom % D |
Safety and Handling
GHS H Statement
Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause drowsiness or dizziness. Suspected of damaging the unborn child.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Warning
missing translation for 'einecsNumber' : 212-742-4
- Chloroform-d
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362-Take off contaminated clothing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.