Learn More
CHLOROFORM HPLC 2.5L
Brand: Fisher Chemical C/4966/17CN
26.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
CHLOROFORM HPLC 2.5L
Additional Details : CAS Number : 67-66-3 Weight : 3.70000kg Transport : UN number : 1888 Chem class : 6.1 Pack group : III