Learn More
Chloroform, HPLC Grade, 99.5+% min, Thermo Scientific Chemicals
£65.30 - £189.00
Chemical Identifiers
CAS | 67-66-3 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 119.37 |
MDL Number | MFCD00000826 |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
PubChem CID | 6212 |
ChEBI | CHEBI:35255 |
SMILES | ClC(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11398187
|
Thermo Scientific Alfa Aesar
022920.K2 |
1 L |
£65.30
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11378187
|
Thermo Scientific Alfa Aesar
022920.M1 |
4 x 1 L |
£189.00
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11388187
|
Thermo Scientific Alfa Aesar
022920.K7 |
4 L | |||||||
Description
Chloroform is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67-66-3 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
ClC(Cl)Cl |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
CHEBI:35255 |
Specifications
°C to 63°C | |
Colorless | |
1.492 g/cm3 at 20°C | |
Characteristic | |
1.446 | |
CHCl3 | |
210 hPa (158mm Hg) at 20°C | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
chloroform | |
6212 | |
119.38 | |
Chloroform |
67-66-3 | |
Colorless liquid | |
60.5°C to 61.5°C | |
Liquid | |
>99.5% | |
MFCD00000826 | |
1731042 | |
Miscible with diethyl ether,oils,ligroin,alcohol,carbon tetrachloride and carbon disulfide. | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 | |
HPLC |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H331-H335-H336-H351-H361d-H373
missing translation for 'dotInformation' : DOT Class: 6.1; Packing Group: III
missing translation for 'einecsNumber' : 200-663-8
missing translation for 'rtecsNumber' : FS9100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Chloroform, HPLC Grade
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : chloroform, 1-pentane
RUO – Research Use Only