missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, HPLC Grade, 99.5+% min, stab. with amylene, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
Description
Chloroform is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | °C to 63°C |
Color | Colorless |
Density | 1.492 g/cm3 at 20°C at 20°C |
Boiling Point | 60.5°C to 61.5°C |
Quantity | 1 L |
Ignition Point | 982°C (1800°F) |
Odor | Characteristic |
Physical Form | Liquid |
Refractive Index | 1.446 |
Assay Percent Range | >99.5% |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Chloroform, HPLC Grade
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : Chloroform, 1-Pentene
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'