Learn More
Chloroform/isoamyl alcohol 24:1(v/v), for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 327155000
158.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For molecular biology
Specifications
Chloroform/isoamyl alcohol 24:1(v/v) | |
DNase, RNase and Protease Free | |
1.4530g/mL | |
500 mL | |
Solution | |
CHCl3 | |
0.56 mPa.s (20°C) | |
Solubility in water: 8g/L (20°C). | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 | |
Molecular Biology |
-63.0°C | |
67-66-3, 123-51-3 | |
62.0°C | |
Glass Bottle | |
1.453 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
chloroform | |
6212 | |
>99.9% (chloroform), ≥99.0% (isoamyl alcohol) (by GC) |
Safety and Handling
- Chloroform/isoamyl alcohol 24:1
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H331-Toxic if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only