missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, J.T. Baker™
221.35 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
4 L
Specifications
Color | Colorless |
Density | 1.483g/cm3 |
Quantity | 1 L |
Physical Form | Liquid |
Formula Weight | 119.38 |
Chemical Name or Material | Chloroform |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Signal Word
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P281-Use personal protective equipment as required.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'