missing translation for 'onlineSavingsMsg'
Learn More

Chloroform Molecular Biology MP Biomedicals

£42.50 - £134.00

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11426461
View Documents
MP Biomedicals
0219400225
25 mL
£42.50
25mL
Please to purchase this item. Need a web account? Register with us today!
11436461
View Documents
MP Biomedicals
0219400280
100 mL
£95.20
100mL
Please to purchase this item. Need a web account? Register with us today!
11446461
View Documents
MP Biomedicals
0219400290
0.5 L
£123.00
500mL
Please to purchase this item. Need a web account? Register with us today!
11456461
View Documents
MP Biomedicals
0219400291
1 L
£134.00
1000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

67-66-3
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
ClC(Cl)Cl
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255

Specifications

67-66-3
CHCl3
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
ClC(Cl)Cl
119.37
CHEBI:35255
Molecular Biology
4.36 (air = 1)
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
119.38
Chloroform
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Chloroform, Molecular Biology Reagent

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : chloroform

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.