missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, ReagentPlus™, ≥99.8%, Honeywell™

Reagent Grade, contains 0.5-1.0% ethanol as stabilizer,≥99.8%

£46.15 - £770.65

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15621290
View Documents Promotion Details
Honeywell Research Chemicals
132950-2.5L
2.5 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15601290
View Documents Promotion Details
Honeywell Research Chemicals
132950-10L
10 L Stainless Steel Can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15631290
View Documents Promotion Details
Honeywell Research Chemicals
132950-25L
25 L Stainless Steel Can
£770.65
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

67-66-3
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
trichloromethane
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
ClC(Cl)Cl

Specifications

-63°C
1.492 g/mL at 25°C, 1.48 g/mL at 25°C
Liquid
CHCl3
160 mmHg (20°C)
1731042
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane
6212
119.38g/mol
ReagentPlus™
67-66-3
60°C to 62°C
n20/D 1.445(lit.)
MFCD00000826
UN1888
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
ClC(Cl)Cl
119.37
CHEBI:35255
≥99.8%
Chloroform
Videos
Safety and Handling

Safety and Handling

P260-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338-P403 + P233

H302-H315-H319-H331-H336-H351-H361d-H372

missing translation for 'dotInformation' : 6.1/PGIII

missing translation for 'einecsNumber' : 200-663-8

Product Identifier
  • Chloroform

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications