Learn More
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -63.0°C |
Absorbance | 0.009 max. at 300nm, 0.046 max. at 265nm, 0.301 max. at 250nm, 0.699 max. at 245nm, 1 cm cell vs water, 1 cm cell vs water at 0nm |
Color | White |
Density | 1.4800g/mL |
Boiling Point | 61.0°C |
Quantity | 2.5 L |
Packaging | Glass Bottle |
Physical Form | Crystals |
Refractive Index | 1.4443 to 1.4463 |
Assay Percent Range | 99+% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466906 | Certificate of Analysis | 31/01/2025 | 390760010,390760025 |
A0465891 | Certificate of Analysis | 07/11/2024 | 390760010,390760025 |
A0463830 | Certificate of Analysis | 02/08/2024 | 390760010,390760025 |
A0459419 | Certificate of Analysis | 11/06/2024 | 390760010,390760025 |
A0455941 | Certificate of Analysis | 22/11/2023 | 390760010,390760025 |
Safety and Handling


- Chloroform, stabilized with ethanol
- Danger
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- H302-Harmful if swallowed.
- H331-Toxic if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H336-May cause drowsiness or dizziness.
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : Chloroform, Ethyl alcohol
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
55Hours
13Minutes
8Seconds
32New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.