missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorogenic acid, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Chlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.
Solubility
Soluble in ethanol and acetone.
Notes
Incompatible with strong oxidizing agents.
Chlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.
Solubility
Soluble in ethanol and acetone.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | 210°C (decomposition) |
Quantity | 1 g |
Merck Index | 14,2142 |
Solubility Information | Soluble in ethanol and acetone. |
Formula Weight | 354.31 |
Physical Form | Powder |
Chemical Name or Material | Chlorogenic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'