Learn More
(Chloromethyl)trimethylsilane, 98%, Thermo Scientific Chemicals
£29.90 - £427.00
Chemical Identifiers
CAS | 2344-80-1 |
---|---|
Molecular Formula | C4H11ClSi |
Molecular Weight (g/mol) | 122.667 |
MDL Number | MFCD00000878 |
InChI Key | OOCUOKHIVGWCTJ-UHFFFAOYSA-N |
Synonym | chloromethyl trimethylsilane, silane, chloromethyl trimethyl, trimethylsilyl methyl chloride, chloromethyl trimethyl silane, ch3 3sich2cl, tmcms, pubchem13668, trimethylchloromethylsilane |
PubChem CID | 75361 |
IUPAC Name | chloromethyl(trimethyl)silane |
SMILES | C[Si](C)(C)CCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11432487
|
Thermo Scientific Alfa Aesar
A14303.09 |
10 g |
£29.90
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11442487
|
Thermo Scientific Alfa Aesar
A14303.18 |
50 g |
£114.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11456837
|
Thermo Scientific Alfa Aesar
A14303.30 |
250 g |
£427.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(Chloromethyl)trimethylsilane is used for Wittig reaction and building blocks.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(Chloromethyl)trimethylsilane is used for Wittig reaction and building blocks.
Solubility
Soluble in water (0.4g/L at 20°C).
Notes
Store in cool, well ventilated area. Keep container tightly closed. Moisture sensitive. Store under argon.
Chemical Identifiers
2344-80-1 | |
122.667 | |
OOCUOKHIVGWCTJ-UHFFFAOYSA-N | |
75361 | |
C[Si](C)(C)CCl |
C4H11ClSi | |
MFCD00000878 | |
chloromethyl trimethylsilane, silane, chloromethyl trimethyl, trimethylsilyl methyl chloride, chloromethyl trimethyl silane, ch3 3sich2cl, tmcms, pubchem13668, trimethylchloromethylsilane | |
chloromethyl(trimethyl)silane |
Specifications
97°C to 98°C | |
C4H11ClSi | |
UN1993 | |
−2°C (28°F) | |
Soluble in water (0.4g/L at 20°C). | |
C[Si](C)(C)CCl | |
122.667 | |
122.67 | |
Odorless | |
Moisture sensitive | |
(Chloromethyl)trimethylsilane |
2344-80-1 | |
MFCD00000878 | |
906705 | |
chloromethyl trimethylsilane, silane, chloromethyl trimethyl, trimethylsilyl methyl chloride, chloromethyl trimethyl silane, ch3 3sich2cl, tmcms, pubchem13668, trimethylchloromethylsilane | |
OOCUOKHIVGWCTJ-UHFFFAOYSA-N | |
chloromethyl(trimethyl)silane | |
75361 | |
98% | |
1.418 | |
0.886 |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 219-058-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only