missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Chloromethylene)dimethylammonium chloride, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
Specifications
Specifications
CAS Min % | 95.0 |
CAS Max % | 100.0 |
Melting Point | 139°C |
Color | Brown to Brown-Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (Argentometry) |
Packaging | Glass bottle |
Linear Formula | ClCH=N(CH3)2Cl |
Quantity | 25 g |
Fieser | 04,186; 05,251; 06,220; 07,422; 08,186; 09,514; 10,457; 12,564; 13,341 |
Show More |
Safety and Handling
Product Identifier
- (Chloromethylene)dimethylammonium chloride
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H360D-May damage the unborn child.
- H302-Harmful if swallowed.
- EUH014-Reacts violently with water.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'