missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethylene)dimethylammonium Chloride, 96%

Product Code. 11401207
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11401207

Brand: Thermo Scientific Alfa Aesar B24172.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3724-43-4 | C3H7Cl2N | 127.996 g/mol

(Chloromethylene)dimethylammonium chloride is used in the synthesis of beta-lactams, n-vinyl substituted indoles and fused 2-pyridone rings and 2-azetidinones. It is used as a formylating agent and used in the preparation of anti-poxvirus agents. It is used in Staudinger reaction and Hundsdiecker-barton iododecarboxylation reaction. It is widely used for studying organic photosensitizers, which finds application in dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Chloromethylene)dimethylammonium chloride is used in the synthesis of beta-lactams, n-vinyl substituted indoles and fused 2-pyridone rings and 2-azetidinones. It is used as a formylating agent and used in the preparation of anti-poxvirus agents. It is used in Staudinger reaction and Hundsdiecker-barton iododecarboxylation reaction. It is widely used for studying organic photosensitizers, which finds application in dye-sensitized solar cells.

Solubility
Soluble in water.

Notes
Moisture sensitive. It may decompose in moist air/ water. Incompatible with bases, strong oxidizing agents, amines, metals and butyl rubber.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3724-43-4
Molecular Formula C3H7Cl2N
MDL Number MFCD00011868
InChI Key QQVDYSUDFZZPSU-UHFFFAOYSA-M
PubChem CID 77311
IUPAC Name chloromethylidene(dimethyl)azanium;chloride

Specifications

Melting Point 130°C (decomposition)
Odor Acrid
Quantity 25 g
UN Number 3261
Beilstein 3566322
Sensitivity Moisture sensitive
Synonym arnold's reagent, vilsmeier reagent, chloromethylene dimethyliminium chloride, methanaminium, n-chloromethylene-n-methyl-, chloride, chloromethylene dimethylammonium chloride, n-chloromethylidene-n-methylmethanaminium chloride, chloromethylenedimethylammonium chloride, chloromethylidene dimethylazanium chloride, n-chloromethylene-n,n-dimethyl ammonium chloride
Solubility Information Soluble in water.
SMILES C[N+](=CCl)C.[Cl-]
Molecular Weight (g/mol) 127.996
Formula Weight 128
Percent Purity 96%
Chemical Name or Material (Chloromethylene)dimethylammonium Chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (Chloromethylene)dimethylammonium chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H360D-May damage the unborn child.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : (Chloromethylene)dimethylammonium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.