missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Chlorophenol Red
Click to view available options
Quantity:
5 g
25 g
Description
Used to detect chlorine dioxide in drinking water. Transition Interval: From pH 4.8 (yellow) to pH 6.7 (violet). Also used in transport studies of goldfish renal tubules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used to detect chlorine dioxide in drinking water. Transition Interval: From pH 4.8 (yellow) to pH 6.7 (violet). Also used in transport studies of goldfish renal tubules.
Solubility
Soluble in water (30 mg/ml), ethanol (80 mg/ml), and methyl cellusolve (50 mg/ml).
Notes
Transition interval: pH 4.8 (yellow) to 6.4 (red)
Used to detect chlorine dioxide in drinking water. Transition Interval: From pH 4.8 (yellow) to pH 6.7 (violet). Also used in transport studies of goldfish renal tubules.
Solubility
Soluble in water (30 mg/ml), ethanol (80 mg/ml), and methyl cellusolve (50 mg/ml).
Notes
Transition interval: pH 4.8 (yellow) to 6.4 (red)
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 5 g |
Beilstein | 354053 |
Solubility Information | Soluble in water (30mg/ml),ethanol (80mg/ml),and methyl cellusolve (50mg/ml). |
Formula Weight | 432.28 |
Chemical Name or Material | Chlorophenol Red |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'