Learn More
Chloroquine diphosphate salt, 98%, Thermo Scientific Chemicals
Chloroquine diphosphate salt, CAS # 50-63-5, is a member of the quinolone family that is considered to be a DNA intercalator. It shows anti-malarial, anti-inflammatory, and anti-filaricides activities.
£92.06 - £195.98
Chemical Identifiers
CAS | 50-63-5 |
---|---|
Molecular Formula | C18H32ClN3O8P2 |
Molecular Weight (g/mol) | 515.86 |
MDL Number | MFCD00069852 |
InChI Key | QKICWELGRMTQCR-UHFFFAOYNA-N |
Synonym | chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquineShow More |
PubChem CID | 64927 |
SMILES | OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15456399
|
Thermo Scientific Alfa Aesar
J64459.14 |
25 g |
£92.06
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15466399
|
Thermo Scientific Alfa Aesar
J64459.22 |
100 g |
£195.98
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chloroquine diphosphate salt is used to study the role of endosomal acidification in cellular processes, such as the signaling of intracellular TLRs. It can be used as DNA intercalator & to dissociate antigen antibody complexes without denaturing red blood cell antigens. Chloroquine showed very high antiviral activity against NiV but very little activity against the other viruses at concentrations lower than 20 μM.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Chloroquine diphosphate salt is a DNA intercalator that shows anti-malarial and anti-inflammatory activities.
- It has been described that this compound interferes with the biosynthesis of nucleic acids, suggesting a cellular mechanism of action
Applications
- Chloroquine diphosphate salt is used as a DNA intercalator to dissociate antigen-antibody complexes without denaturing red blood cell antigens
- This compound shows antimalarial activity against Plasmodium falciparum, except against chloroquine-resistant strains. It has been used in in vitro antiplasmodial assays. In parasites, the mechanism of action remains unclear, but this compound can inhibit the parasitic enzyme heme polymerase that converts the toxic heme into non-toxic hemozoin, resulting in the accumulation of toxic heme within the parasite.
- It shows very high anti-viral activity against NiV but lower activity against other viruses at concentrations lower than 20 μM
- Chloroquine diphosphate salt is used to study the role of endosomal acidification in cellular processes, such as the signaling of intracellular TLRs
- Chloroquine diphosphate salt has been used in autophagy inhibition studies
Chemical Identifiers
50-63-5 | |
515.86 | |
QKICWELGRMTQCR-UHFFFAOYNA-N | |
64927 |
C18H32ClN3O8P2 | |
MFCD00069852 | |
chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquineShow More | |
OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1 |
Specifications
50-63-5 | |
White | |
MFCD00069852 | |
14,2163 | |
Soluble in water | |
OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1 | |
515.86 | |
515.87 | |
Powder |
213°C to 216°C | |
C18H32ClN3O8P2 | |
4223142 | |
chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquine | |
QKICWELGRMTQCR-UHFFFAOYNA-N | |
4-N-(7-chloroquinolin-4-yl)-1-N,1-N-diethylpentane-1,4-diamine;phosphoric acid | |
64927 | |
98% | |
Chloroquine diphosphate salt |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 200-055-2
missing translation for 'rtecsNumber' : VB2450000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.